Assay Details
Gene Symbol: SRD5A1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_023625866.1 | XP_023481634.1 | 632 | 82 | ||
XM_023625867.1 | XP_023481635.1 | 633 | 82 |

More Information
Set Membership: |
![]() ![]() ![]() |
