Assay Details
Gene Symbol: CST7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005604430.3 | XP_005604487.2 | 365 | 82 | ||
XM_005604431.3 | XP_005604488.1 | 372 | 82 | |||
XM_023625989.1 | XP_023481757.1 | 333 | 82 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
