Assay Details
Gene Symbol: IRAG2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_023643246.1 | XP_023499014.1 | 613 | 75 | ||
GenBank mRNA | JL640328.1 | - | 4 - 5 | 456 | 75 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
