Assay Details
Gene Symbol: IRAG2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_023643246.1 | XP_023499014.1 | 1116 | 76 | ||
GenBank mRNA | JL618101.1 | - | 4 - 5 | 321 | 76 |

More Information
Set Membership: |
![]() ![]() ![]() |
