Assay Details
Gene Symbol: LMNTD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_014740651.2 | XP_014596137.2 | 263 | 89 | ||
XM_023643252.1 | XP_023499020.1 | 1492 | 89 | |||
XM_023643253.1 | XP_023499021.1 | 857 | 89 | |||
XM_023643254.1 | XP_023499022.1 | 869 | 89 | |||
XM_023643255.1 | XP_023499023.1 | 1429 | 89 | |||
XM_023643256.1 | XP_023499024.1 | 793 | 89 | |||
XM_023643257.1 | XP_023499025.1 | 917 | 89 | |||
XM_023643258.1 | XP_023499026.1 | 1029 | 89 | |||
XM_023643259.1 | XP_023499027.1 | 4133 | 89 | |||
XM_023643260.1 | XP_023499028.1 | 1492 | 89 | |||
XM_023643261.1 | XP_023499029.1 | 1493 | 89 | |||
XM_023643262.1 | XP_023499030.1 | 732 | 89 | |||
XM_023643263.1 | XP_023499031.1 | 471 | 89 | |||
XM_023643264.1 | XP_023499032.1 | 2741 | 89 | |||
XM_023643265.1 | XP_023499033.1 | 366 | 89 | |||
XM_023643266.1 | XP_023499034.1 | 1492 | 89 | |||
XM_023643267.1 | XP_023499035.1 | 1369 | 89 | |||
XM_023643268.1 | XP_023499036.1 | 278 | 89 | |||
XM_023643269.1 | XP_023499037.1 | 251 | 89 | |||
XM_023643270.1 | XP_023499038.1 | 281 | 89 | |||
XM_023643271.1 | XP_023499039.1 | 272 | 89 |

More Information
Set Membership: |
![]() ![]() ![]() |
