Assay Details
Gene Symbol: SSPN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001498823.5 | XP_001498873.2 | 344 | 100 | ||
XM_014740544.2 | XP_014596030.1 | 402 | 100 | |||
XM_023644094.1 | XP_023499862.1 | 436 | 100 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
