Assay Details
Gene Symbol: INTS13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001502728.6 | XP_001502778.3 | 1431 | 66 | ||
XM_014740702.2 | XP_014596188.2 | 1431 | 66 | |||
XM_023643277.1 | XP_023499045.1 | 981 | 66 |

More Information
Set Membership: |
![]() ![]() ![]() |
