Assay Details
Gene Symbol: FGFR1OP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001502758.4 | XP_001502808.1 | 786 | 77 | ||
XM_023643278.1 | XP_023499046.1 | 906 | 77 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
