Assay Details
Gene Symbol: FAM186B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001492150.4 | XP_001492200.2 | 3076 | 64 | ||
XM_023643502.1 | XP_023499270.1 | 2779 | 64 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
