Assay Details
Gene Symbol: TMBIM6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001504223.4 | XP_001504273.1 | 350 | 59 | ||
XM_023643512.1 | XP_023499280.1 | 217 | 59 | |||
GenBank mRNA | JL616479.2 | - | 3 - 4 | 255 | 59 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
