Assay Details
Gene Symbol: ATF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001492609.5 | XP_001492659.3 | 583 | 69 | ||
XM_014740977.2 | XP_014596463.1 | 704 | 69 | |||
XM_014740978.2 | XP_014596464.1 | 481 | 69 | |||
XM_023643554.1 | XP_023499322.1 | 688 | 69 | |||
XM_023643555.1 | XP_023499323.1 | 589 | 69 | |||
XM_023643556.1 | XP_023499324.1 | 472 | 69 | |||
XM_023643557.1 | XP_023499325.1 | 567 | 69 | |||
XM_023643558.1 | XP_023499326.1 | 567 | 69 | |||
XM_023643559.1 | XP_023499327.1 | 469 | 69 | |||
XM_023643560.1 | XP_023499328.1 | 351 | 69 | |||
XM_023643561.1 | XP_023499329.1 | 446 | 69 |

More Information
Set Membership: |
![]() ![]() ![]() |
