Assay Details
Gene Symbol: NAPB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_023625998.1 | XP_023481766.1 | 265 | 66 | ||
XM_023625999.1 | XP_023481767.1 | 233 | 66 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_023625999.1 transcripts.
Set Membership: |
![]() ![]() ![]() |
