Assay Details
Gene Symbol: DTD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_023626045.1 | XP_023481813.1 | 546 | 65 | ||
XM_023626046.1 | XP_023481814.1 | 548 | 65 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
