Assay Details
Gene Symbol: RBBP9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001490566.4 | XP_001490616.1 | 288 | 77 | ||
XM_023626049.1 | XP_023481817.1 | 441 | 77 |

More Information
Set Membership: |
![]() ![]() ![]() |
