Assay Details
Gene Symbol: KAT14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001493916.6 | XP_001493966.2 | 1698 | 82 | ||
XM_023626068.1 | XP_023481836.1 | 1402 | 82 |

More Information
Set Membership: |
![]() ![]() ![]() |
