Assay Details
Gene Symbol: PAK5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_014735031.2 | XP_014590517.1 | 1326 | 58 | ||
XM_023626124.1 | XP_023481892.1 | 1425 | 58 | |||
XM_023626125.1 | XP_023481893.1 | 1504 | 58 |

More Information
Set Membership: |
![]() ![]() ![]() |
