Assay Details
Gene Symbol: LAMP5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001493398.5 | XP_001493448.1 | 554 | 74 | ||
XM_003363879.4 | XP_003363927.1 | 555 | 74 |

More Information
Set Membership: |
![]() ![]() ![]() |
