Assay Details
Gene Symbol: PCNA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001494721.5 | XP_001494771.1 | 435 | 78 | ||
GenBank mRNA | JL616384.1 | - | 1 - 2 | 385 | 78 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
