Assay Details
Gene Symbol: GFRA4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_023626198.1 | XP_023481966.1 | 1122 | 75 | ||
XM_023626199.1 | XP_023481967.1 | 1119 | 75 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
