Assay Details
Gene Symbol: FKBP1A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_023626260.1 | XP_023482028.1 | 258 | 87 | ||
XM_023626261.1 | XP_023482029.1 | 346 | 87 |

More Information
Set Membership: |
![]() ![]() ![]() |
