Assay Details
Gene Symbol: ANGPT4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001497777.4 | XP_001497827.1 | 1437 | 63 | ||
XM_005604539.3 | XP_005604596.1 | 1438 | 63 | |||
XM_014735083.2 | XP_014590569.1 | 1438 | 63 |

More Information
Set Membership: |
![]() ![]() ![]() |
