Assay Details
Gene Symbol: TRIB3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001499281.4 | XP_001499331.1 | 1567 | 94 | ||
XM_023626279.1 | XP_023482047.1 | 1737 | 94 |

More Information
Set Membership: |
![]() ![]() ![]() |
