Assay Details
Gene Symbol: CCM2L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_023626298.1 | XP_023482066.1 | 259 | 59 | ||
XM_023626299.1 | XP_023482067.1 | 259 | 59 | |||
XM_023626300.1 | XP_023482068.1 | 259 | 59 | |||
XM_023626301.1 | XP_023482069.1 | 259 | 59 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
