Assay Details
Gene Symbol: ASXL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_023626308.1 | XP_023482076.1 | 539 | 68 | ||
XM_023626309.1 | XP_023482077.1 | 542 | 68 | |||
XM_023626310.1 | XP_023482078.1 | 556 | 68 | |||
XM_023626312.1 | XP_023482080.1 | 563 | 68 |

More Information
Set Membership: |
![]() ![]() ![]() |
