Assay Details
Gene Symbol: EIF6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001501536.4 | XP_001501586.1 | 346 | 79 | ||
GenBank mRNA | JL615902.1 | - | 3 - 4 | 299 | 79 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
