Assay Details
Gene Symbol: TTI1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001502393.6 | XP_001502443.3 | 2457 | 55 | ||
XM_023626541.1 | XP_023482309.1 | 2422 | 55 |

More Information
Set Membership: |
![]() ![]() ![]() |
