Assay Details
Gene Symbol: TP53TG5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001500634.4 | XP_001500684.1 | 229 | 86 | ||
XM_023626624.1 | XP_023482392.1 | 353 | 86 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: TP53TG5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001500634.4 | XP_001500684.1 | 229 | 86 | ||
XM_023626624.1 | XP_023482392.1 | 353 | 86 |