Assay Details
Gene Symbol: UBE2V1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_014735208.2 | XP_014590694.1 | 268 | 108 | ||
XM_023626733.1 | XP_023482501.1 | 396 | 108 | |||
XM_023626734.1 | XP_023482502.1 | 293 | 108 | |||
XM_023626735.1 | XP_023482503.1 | 442 | 108 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
