Assay Details
Gene Symbol: RIPOR3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001487915.5 | XP_001487965.2 | 3546 | 55 | ||
XM_023626739.1 | XP_023482507.1 | 3688 | 55 | |||
XM_023626740.1 | XP_023482508.1 | 3685 | 55 | |||
XM_023626741.1 | XP_023482509.1 | 5689 | 55 | |||
XM_023626742.1 | XP_023482510.1 | 2995 | 55 | |||
XM_023626743.1 | XP_023482511.1 | 3549 | 55 |

More Information
Set Membership: |
![]() ![]() ![]() |
