Assay Details
Gene Symbol: ADNP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_014735282.2 | XP_014590768.1 | 1823 | 111 | ||
XM_023626746.1 | XP_023482514.1 | 456 | 111 | |||
XM_023626747.1 | XP_023482515.1 | 494 | 111 | |||
XM_023626748.1 | XP_023482516.1 | 280 | 111 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
