Assay Details
Gene Symbol: AURKA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001489270.6 | XP_001489320.1 | 548 | 69 | ||
XM_005604797.3 | XP_005604854.1 | 548 | 69 | |||
XM_023626777.1 | XP_023482545.1 | 1220 | 69 | |||
XM_023626778.1 | XP_023482546.1 | 548 | 69 | |||
XM_023626779.1 | XP_023482547.1 | 548 | 69 | |||
XM_023626780.1 | XP_023482548.1 | 548 | 69 | |||
XM_023626781.1 | XP_023482549.1 | 548 | 69 |

More Information
Set Membership: |
![]() ![]() ![]() |
