Assay Details
Gene Symbol: CASS4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_014735296.2 | XP_014590782.2 | 383 | 75 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_014735296.2 transcripts.
Set Membership: |
![]() ![]() |
