Assay Details
Gene Symbol: CASS4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005604798.3 | XP_005604855.2 | 1430 | 100 | ||
XM_014735296.2 | XP_014590782.2 | 919 | 100 | |||
XM_023626783.1 | XP_023482551.1 | 5355 | 100 |
