Assay Details
Gene Symbol: PRELID3B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_023626830.1 | XP_023482598.1 | 584 | 83 | ||
XM_023626831.1 | XP_023482599.1 | 587 | 83 | |||
GenBank mRNA | JL616367.2 | - | 4 - 5 | 422 | 83 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
