Assay Details
Gene Symbol: SS18L1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_023626860.1 | XP_023482628.1 | 1008 | 56 | ||
XM_023626861.1 | XP_023482629.1 | 3959 | 56 | |||
XM_023626862.1 | XP_023482630.1 | 4668 | 56 | |||
XM_023626863.1 | XP_023482631.1 | 1738 | 56 | |||
XM_023626864.1 | XP_023482632.1 | 1008 | 56 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
