Assay Details
Gene Symbol: RBBP8NL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_014735319.2 | XP_014590805.1 | 741 | 61 | ||
XM_014735320.2 | XP_014590806.1 | 740 | 61 | |||
XM_014735321.2 | XP_014590807.1 | 742 | 61 |

More Information
Set Membership: |
![]() ![]() ![]() |
