Assay Details
Gene Symbol: CEMIP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005604959.3 | XP_005605016.1 | 3867 | 55 | ||
XM_005604960.3 | XP_005605017.1 | 3784 | 55 |

More Information
Set Membership: |
![]() ![]() ![]() |
