Assay Details
Gene Symbol: AK3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_023627306.1 | XP_023483074.1 | 599 | 75 | ||
XM_023627307.1 | XP_023483075.1 | 599 | 75 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
