Assay Details
Gene Symbol: RIC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_023627318.1 | XP_023483086.1 | 3032 | 60 | ||
XM_023627319.1 | XP_023483087.1 | 3032 | 60 | |||
XM_023627320.1 | XP_023483088.1 | 3029 | 60 | |||
XM_023627321.1 | XP_023483089.1 | 2879 | 60 | |||
XM_023627322.1 | XP_023483090.1 | 2975 | 60 | |||
XM_023627323.1 | XP_023483091.1 | 2991 | 60 |

More Information
Set Membership: |
![]() ![]() ![]() |
