Assay Details
Gene Symbol: SMU1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_023627466.1 | XP_023483234.1 | 617 | 81 | ||
GenBank mRNA | JL615963.1 | - | 4 - 5 | 508 | 81 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
