Assay Details
Gene Symbol: UBE2R2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_023627481.1 | XP_023483249.1 | 743 | 83 | ||
XM_023627482.1 | XP_023483250.1 | 650 | 83 | |||
XM_023627483.1 | XP_023483251.1 | 411 | 83 | |||
XM_023627484.1 | XP_023483252.1 | 318 | 83 |

More Information
Set Membership: |
![]() ![]() ![]() |
