Assay Details
Gene Symbol: DCAF12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001499560.6 | XP_001499610.1 | 907 | 64 | ||
XM_005605060.3 | XP_005605117.1 | 720 | 64 |

More Information
Set Membership: |
![]() ![]() ![]() |
