Assay Details
Gene Symbol: UBAP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001499590.5 | XP_001499640.1 | 1328 | 74 | ||
XM_005605061.3 | XP_005605118.1 | 1309 | 74 | |||
XM_014735513.2 | XP_014590999.1 | 1201 | 74 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
