Assay Details
Gene Symbol: CENPP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005605114.3 | XP_005605171.1 | 1178 | 77 | ||
XM_023627640.1 | XP_023483408.1 | 791 | 77 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
