Assay Details
Gene Symbol: ECM2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001491657.6 | XP_001491707.4 | 1185 | 65 | ||
XM_005605111.3 | XP_005605168.1 | 1214 | 65 | |||
XM_005605112.3 | XP_005605169.1 | 2497 | 65 | |||
XM_014735539.2 | XP_014591025.1 | 1263 | 65 | |||
XM_023627632.1 | XP_023483400.1 | 1257 | 65 |

More Information
Set Membership: |
![]() ![]() ![]() |
