Assay Details
Gene Symbol: GNPNAT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_023627743.1 | XP_023483511.1 | 549 | 84 | ||
GenBank mRNA | JL616006.2 | - | 5 - 6 | 450 | 84 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
