Assay Details
Gene Symbol: CDKN3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001490077.6 | XP_001490127.3 | 689 | 68 | ||
XM_005605189.3 | XP_005605246.1 | 671 | 68 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
