Assay Details
Gene Symbol: PELI2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_023627785.1 | XP_023483553.1 | 479 | 70 | ||
XM_023627786.1 | XP_023483554.1 | 248 | 70 |

More Information
Set Membership: |
![]() ![]() ![]() |
