Assay Details
Gene Symbol: TIMM9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005605209.3 | XP_005605266.1 | 309 | 81 | ||
XM_014735621.2 | XP_014591107.1 | 352 | 81 |

More Information
Set Membership: |
![]() ![]() ![]() |
