Assay Details
Gene Symbol: RDH11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001494572.5 | XP_001494622.4 | 541 | 78 | ||
XM_014735661.2 | XP_014591147.1 | 499 | 78 |

More Information
Set Membership: |
![]() ![]() ![]() |
